Anti-C/EBP b (2 isoforms C/EBP b, LAP) Mouse Monoclonal Antibody [clone: A16]

Supplier: Biolegend
626001 626002
CA626001-BLEA 161.18 CAD
CA626001-BL CA626002-BL
Anti-C/EBP b (2 isoforms C/EBP b, LAP) Mouse Monoclonal Antibody [clone: A16]
Antibodies
Purified anti-C/EBP β (2 isoforms C/EBP β, LAP) [A16]; Isotype: Mouse IgG; Reactivity: Mouse (recognizes 35 and 32 kD LAP forms); Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: C/EBP b (2 isoforms C/EBP b, LAP)
Clonality: Monoclonal
Clone: A16
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR