Anti-Alix Mouse Monoclonal Antibody [clone: 3A9]

Supplier: Biolegend
634501 634502
CA634501-BLEA 178.73 CAD
CA634501-BL CA634502-BL
Anti-Alix Mouse Monoclonal Antibody [clone: 3A9]
Antibodies
Purified anti-Alix [3A9]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, and Frog; Apps: WB, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 1 μg/ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Alix
Clonality: Monoclonal
Clone: 3A9
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse;and Frog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR