Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: RB6-8C5]

Supplier: Biolegend
108427 108428
CA108427-BLEA 164.37 CAD
CA108427-BL CA108428-BL
Anti-Ly-6G/Ly-6C (Gr-1) Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: RB6-8C5]
Antibodies
PerCP/Cy5.5 anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6G/Ly-6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR