Anti-CD69 Armenian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: H1.2F3]

Supplier: Biolegend
104521 104522
CA104521-BLEA 188.3 CAD
CA104521-BL CA104522-BL
Anti-CD69 Armenian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: H1.2F3]
Antibodies
PerCP/Cy5.5 anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol CD69
Environmentally Preferable
Antigen synonyms EA1|MLR3|gp34/28|AIM|Very Early Activation Antigen (VEA)
Antibody type Primary
Clonality Monoclonal
Conjugation PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Clone H1.2F3
Reactivity Mouse
Host Armenian hamster
Gene ID 12515
Isotype IgG
Flow cytometry Yes
Immunogen Mouse dendritic epidermal T cell line Y245
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
Concentration 0.20 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR