Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: 145-2C11]

Supplier: Biolegend
100327 100328
CA100327-BLEA 130.86 CAD
CA100327-BL CA100328-BL
Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: 145-2C11]
Antibodies
PerCP/Cy5.5 anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR