Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: TER-119]

Supplier: Biolegend
116221 116222
CA116221-BLEA 161.18 CAD
CA116221-BL CA116222-BL
Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: TER-119]
Antibodies
PE/Cy7 anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TER-119/Erythroid Cells
Clonality: Monoclonal
Clone: TER-119
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR