Anti-Ly-6A/E (Sca-1) Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: E13-161.7]

Supplier: Biolegend
122513 122514 122514
CA122513-BLEA 161.18 CAD
CA122513-BL CA122514-BL 122514-BL
Anti-Ly-6A/E (Sca-1) Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: E13-161.7]
Antibodies
PE/Cy7 anti-mouse Ly-6A/E (Sca-1) [E13-161.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ly-6A/E (Sca-1)
Clonality: Monoclonal
Clone: E13-161.7
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR