Anti-CD8a Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 53-6.7]

Supplier: Biolegend
100721 100722
CA100721-BLEA 108.51 CAD
CA100721-BL CA100722-BL
Anti-CD8a Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 53-6.7]
Antibodies
PE/Cy7 anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR