Anti-CD45 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 30-F11]

Supplier: Biolegend
103113 103114
CA103113-BLEA 138.83 CAD
CA103113-BL CA103114-BL
Anti-CD45 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 30-F11]
Antibodies
PE/Cy7 anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR