Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: GK1.5]

Supplier: Biolegend
100421 100422
CA100421-BLEA 140.43 CAD
CA100421-BL CA100422-BL
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: GK1.5]
Antibodies
PE/Cy7 anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR