Anti-CD5 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UCHT2]

Supplier: Biolegend
300621 300622
CA300621-BLEA 272.88 CAD
CA300621-BL CA300622-BL
Anti-CD5 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: UCHT2]
Antibodies
PE/Cy7 anti-human CD5 [UCHT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Capuchin Monkey, Common Marmoset, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Capuchin Monkey;Common Marmoset;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR