Anti-CD86 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 24F]

Supplier: Biolegend
200307 200308
CA200307-BLEA 175.54 CAD
CA200307-BL CA200308-BL
Anti-CD86 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 24F]
Antibodies
PE anti-rat CD86 [24F]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 24F
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR