Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TER-119]

Supplier: Biolegend
116207 116208
CA116207-BLEA 116.49 CAD
CA116207-BL CA116208-BL
Anti-TER-119/Erythroid Cells Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TER-119]
Antibodies
PE anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TER-119/Erythroid Cells
Clonality: Monoclonal
Clone: TER-119
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR