Anti-Ig light chain lambda Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RML-42]

Supplier: Biolegend
407307 407308
CA407307-BLEA 175.54 CAD
CA407307-BL CA407308-BL
Anti-Ig light chain lambda Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RML-42]
Antibodies
PE anti-mouse Ig light chain λ [RML-42]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ig light chain lambda
Clonality: Monoclonal
Clone: RML-42
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR