Anti-I-Ab Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AF6-120.1]

Supplier: Biolegend
116407 116408
CA116407-BLEA 194.69 CAD
CA116407-BL CA116408-BL
Anti-I-Ab Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AF6-120.1]
Antibodies
PE anti-mouse I-Ab [AF6-120.1]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, Cross-reacts with H-2 k and H-2 u haplotypes. Does not react with other haplotypes (e.g., d, f, q, r, s).; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: I-Ab
Clonality: Monoclonal
Clone: AF6-120.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG2a, κ
Reactivity: Mouse b haplotype;Cross-reacts with H-2 k and H-2 u haplotypes. Does not react with other haplotypes (e.g.;d;f;q;r;s).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR