Anti-H-2Kb Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AF6-88.5]

Supplier: Biolegend
116507 116508
CA116507-BLEA 194.69 CAD
CA116507-BL CA116508-BL
Anti-H-2Kb Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: AF6-88.5]
Antibodies
PE anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, Does not cross-react with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v); Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb
Clonality: Monoclonal
Clone: AF6-88.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG2a, κ
Reactivity: Mouse b haplotype;Does not cross-react with other haplotypes (e.g.;d;f;j;k;p;q;r;s;u;v)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR