Anti-H-2Dd Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 34-2-12]

Supplier: Biolegend
110607 110608
CA110607-BLEA 205.86 CAD
CA110607-BL CA110608-BL
Anti-H-2Dd Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 34-2-12]
Antibodies
PE anti-mouse H-2Dd [34-2-12]; Isotype: Mouse (C3H/HeJ) IgG2a, κ; Reactivity: Mouse d haplotype, Cross-reacts with the C3H.LG/Ckc. Does not cross react with other haplotypes (e.g., b, f, k, p, q, r, s).; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Dd
Clonality: Monoclonal
Clone: 34-2-12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse (C3H/HeJ) IgG2a, κ
Reactivity: Mouse d haplotype;Cross-reacts with the C3H.LG/Ckc. Does not cross react with other haplotypes (e.g.;b;f;k;p;q;r;s).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR