Anti-H-2Db Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: KH95]

Supplier: Biolegend
111507 111508
CA111507-BLEA 205.86 CAD
CA111507-BL CA111508-BL
Anti-H-2Db Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: KH95]
Antibodies
PE anti-mouse H-2Db [KH95]; Isotype: Mouse (BALB/c) IgG2b, κ; Reactivity: Mouse b haplotype, Does not cross-react with other haplotypes (e.g., a, d, f, k, n, p, q, r, s, u, v); Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Db
Clonality: Monoclonal
Clone: KH95
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG2b, κ
Reactivity: Mouse b haplotype;Does not cross-react with other haplotypes (e.g.;a;d;f;k;n;p;q;r;s;u;v)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR