Anti-H-2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M1/42]

Supplier: Biolegend
125505 125506
CA125505-BLEA 175.54 CAD
CA125505-BL CA125506-BL
Anti-H-2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: M1/42]
Antibodies
PE anti-mouse H-2 [M1/42]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for other applications.

Type: Primary
Antigen: H-2
Clonality: Monoclonal
Clone: M1/42
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR