Anti-FasL (CD178) Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MFL3]

Supplier: Biolegend
106605 106606
CA106605-BLEA 175.54 CAD
CA106605-BL CA106606-BL
Anti-FasL (CD178) Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MFL3]
Antibodies
PE anti-mouse CD178 (FasL) [MFL3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: FasL (CD178)
Clonality: Monoclonal
Clone: MFL3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR