Anti-DO11.10 TCR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: KJ1-26]

Supplier: Biolegend
118507 118508
CA118507-BLEA 137.24 CAD
CA118507-BL CA118508-BL
Anti-DO11.10 TCR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: KJ1-26]
Antibodies
PE anti-mouse TCR DO11.10 [KJ1-26]; Isotype: Mouse IgG2a, κ; Reactivity: mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: DO11.10 TCR
Clonality: Monoclonal
Clone: KJ1-26
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR