Anti-CD90.2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 30-H12]

Supplier: Biolegend
105307 105308
CA105307-BLEA 116.49 CAD
CA105307-BL CA105308-BL
Anti-CD90.2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 30-H12]
Antibodies
PE anti-mouse CD90.2 [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR