Anti-CD51 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMV-7]

Supplier: Biolegend
104105 104106
CA104105-BLEA 156.39 CAD
CA104105-BL CA104106-BL
Anti-CD51 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RMV-7]
Antibodies
PE anti-mouse CD51 [RMV-7]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD51
Clonality: Monoclonal
Clone: RMV-7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR