Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RM4-5]

Supplier: Biolegend
100511 100512
CA100511-BLEA 39.9 CAD
CA100511-BL CA100512-BL
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RM4-5]
Antibodies
PE anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR