Anti-CD31 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 390]

Supplier: Biolegend
102407 102408
CA102407-BLEA 175.54 CAD
CA102407-BL CA102408-BL
Anti-CD31 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 390]
Antibodies
PE anti-mouse CD31 [390]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 390
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol CD31
Environmentally Preferable
Antigen synonyms PECAM-1|gpIIa|EndoCAM
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone 390
Reactivity Mouse
Host Rat
Gene ID 18613
Isotype IgG2a kappa
Flow cytometry Yes
Immunogen C3H/HeJ mouse hematopoietic progenitor cell line 3
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
Concentration 0.20 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR