Anti-CD226 (DNAM-1) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10E5]

Supplier: Biolegend
128805 128806
CA128805-BLEA 194.69 CAD
CA128805-BL CA128806-BL
Anti-CD226 (DNAM-1) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10E5]
Antibodies
PE anti-mouse CD226 (DNAM-1) [10E5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD226 (DNAM-1)
Clonality: Monoclonal
Clone: 10E5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR