Anti-CD132 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TUGm2]

Supplier: Biolegend
132305 132306
CA132305-BLEA 232.99 CAD
CA132305-BL CA132306-BL
Anti-CD132 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TUGm2]
Antibodies
PE anti-mouse CD132 (common γ chain) [TUGm2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD132
Clonality: Monoclonal
Clone: TUGm2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR