Anti-CD122 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TM-beta1]

Supplier: Biolegend
123209 123210
CA123209-BLEA 137.24 CAD
CA123209-BL CA123210-BL
Anti-CD122 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: TM-beta1]
Antibodies
PE anti-mouse CD122 (IL-2Rβ) [TM-β1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD122
Clonality: Monoclonal
Clone: TM-beta1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR