Anti-CD103 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2E7]

Supplier: Biolegend
121405 121406
CA121405-BLEA 116.49 CAD
CA121405-BL CA121406-BL
Anti-CD103 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2E7]
Antibodies
PE anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: 2E7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR