Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: SK7]

Supplier: Biolegend
344805 344806
CA344805-BLEA 167.56 CAD
CA344805-BL CA344806-BL
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: SK7]
Antibodies
PE anti-human CD3 [SK7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsizeMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD3
Environmentally Preferable
Antigen synonyms T3|CD3ε
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone SK7
Reactivity Chimpanzee,Human
Host Mouse
Gene ID 916
Isotype IgG1 kappa
Flow cytometry Yes
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR