Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIB19]

Supplier: Biolegend
302207 302208 302254
CA302207-BLEA 44.68 CAD
CA302207-BL CA302208-BL CA10757-712
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIB19]
Antibodies
PE anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Chimpanzee;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR