Anti-CD178 (Fas-L) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NOK-1]

Supplier: Biolegend
306406 306407
CA306406-BLEA 193.09 CAD
CA306406-BL CA306407-BL
Anti-CD178 (Fas-L) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NOK-1]
Antibodies
PE anti-human CD178 (Fas-L) [NOK-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: CD178 (Fas-L)
Clonality: Monoclonal
Clone: NOK-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR