Anti-TCR-b chain Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: H57-597]

Supplier: Biolegend
109225 109226
CA109225-BLEA 150.01 CAD
CA109225-BL CA109226-BL
Anti-TCR-b chain Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: H57-597]
Antibodies
Pacific Blue™ anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.5 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCR-b chain
Clonality: Monoclonal
Clone: H57-597
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR