Anti-CD86 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GL-1]

Supplier: Biolegend
105021 105022
CA105021-BLEA 161.18 CAD
CA105021-BL CA105022-BL
Anti-CD86 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GL-1]
Antibodies
Pacific Blue™ anti-mouse CD86 [GL-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL-1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR