Anti-CD69 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: H1.2F3]

Supplier: Biolegend
104523 104524
CA104523-BLEA 150.01 CAD
CA104523-BL CA104524-BL
Anti-CD69 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: H1.2F3]
Antibodies
Pacific Blue™ anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR