Anti-CD48 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: HM48-1]

Supplier: Biolegend
103417 103418
CA103417-BLEA 161.18 CAD
CA103417-BL CA103418-BL
Anti-CD48 Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: HM48-1]
Antibodies
Pacific Blue™ anti-mouse CD48 [HM48-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: HM48-1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR