Anti-CD45.2 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 104]

Supplier: Biolegend
109819 109820
CA109819-BLEA 150.01 CAD
CA109819-BL CA109820-BL
Anti-CD45.2 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 104]
Antibodies
Pacific Blue™ anti-mouse CD45.2 [104]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45.2
Clonality: Monoclonal
Clone: 104
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse (SJL) IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR