Anti-CD45 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 30-F11]

Supplier: Biolegend
103125 103126
CA103125-BLEA 150.01 CAD
CA103125-BL CA103126-BL
Anti-CD45 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 30-F11]
Antibodies
Pacific Blue™ anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR