Anti-CD3 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 17A2]

Supplier: Biolegend
100213 100214
CA100213-BLEA 150.01 CAD
CA100213-BL CA100214-BL
Anti-CD3 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 17A2]
Antibodies
Pacific Blue™ anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR