Anti-CD11c Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: N418]

Supplier: Biolegend
117321 117322
CA117321-BLEA 150.01 CAD
CA117321-BL CA117322-BL
Anti-CD11c Armenian Hamster Monoclonal Antibody (PB (Pacific Blue)) [clone: N418]
Antibodies
Pacific Blue™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: PB (Pacific Blue)
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR