Anti-CD117 (c-kit) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 2B8]

Supplier: Biolegend
105819 105820 105820
CA105819-BLEA 161.18 CAD
CA105819-BL CA105820-BL 105820-BL
Anti-CD117 (c-kit) Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 2B8]
Antibodies
Pacific Blue™ anti-mouse CD117 (c-Kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD117 (c-kit)
Clonality: Monoclonal
Clone: 2B8
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol CD117 (c-kit)
Environmentally Preferable
Antigen synonyms KIT|c-KIT|Stem Cell Factor Receptor (SCFR)
Antibody type Primary
Clonality Monoclonal
Conjugation PB (Pacific Blue)
Clone 2B8
Reactivity Mouse
Host Rat
Gene ID 16590
Isotype IgG2b kappa
Flow cytometry Yes
Immunogen Mouse bone marrow mast cells
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR