Anti-HLA-A,B,C Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: W6/32]

Supplier: Biolegend
311417 311418
CA311417-BLEA 197.88 CAD
CA311417-BL CA311418-BL
Anti-HLA-A,B,C Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: W6/32]
Antibodies
Pacific Blue™ anti-human HLA-A,B,C [W6/32]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1..0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: HLA-A,B,C
Clonality: Monoclonal
Clone: W6/32
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cat (Feline);Cattle (Bovine;Cow);Chimpanzee;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR