Anti-CD8a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIT8a]

Supplier: Biolegend
300927 300928
CA300927-BLEA 202.67 CAD
CA300927-BL CA300928-BL
Anti-CD8a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HIT8a]
Antibodies
Pacific Blue™ anti-human CD8a [HIT8a]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Marmoset; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: HIT8a
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR