Anti-CD8a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: RPA-T8]

Supplier: Biolegend
301023 301026 301033
CA301023-BLEA 427.67 CAD
CA301023-BL CA301026-BL CA301033-BL
Anti-CD8a Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: RPA-T8]
Antibodies
Pacific Blue™ anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume. 
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 1.0 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: RPA-T8
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Pigtailed Macaque;Sooty Mangabey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR