Anti-CD86 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: IT2.2]

Supplier: Biolegend
305417 305418 305423
CA305417-BLEA 197.88 CAD
CA305417-BL CA305418-BL CA305423-BL
Anti-CD86 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: IT2.2]
Antibodies
Pacific Blue™ anti-human CD86 [IT2.2]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Common Marmoset, Cotton-topped Tamarin, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per 106 cells in 100 μl volume.
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 0.5 μg per 106 cells in 100 μl volume.
It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: IT2.2
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Common Marmoset;Cotton-topped Tamarin;Chimpanzee;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR