Anti-CD54 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HA58]

Supplier: Biolegend
353109 353110
CA353109-BLEA 234.58 CAD
CA353109-BL CA353110-BL
Anti-CD54 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HA58]
Antibodies
Pacific Blue™ anti-human CD54 [HA58]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: HA58
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR