Anti-CD54 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HCD54]

Supplier: Biolegend
322715 322716
CA322715-BLEA 197.88 CAD
CA322715-BL CA322716-BL
Anti-CD54 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: HCD54]
Antibodies
Pacific Blue™ anti-human CD54 [HCD54]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: HCD54
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR