Anti-CD45RO Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: UCHL1]

Supplier: Biolegend
304215 304216
CA304215-BLEA 197.88 CAD
CA304215-BL CA304216-BL
Anti-CD45RO Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: UCHL1]
Antibodies
Pacific Blue™ anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: UCHL1
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Chimpanzee;Cynomolgus;Common Marmoset
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR