Anti-CD4 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: OKT4]

Supplier: Biolegend
317423 317424 317429
CA317423-BLEA 197.88 CAD
CA317423-BL CA317424-BL CA317429-BL
Anti-CD4 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: OKT4]
Antibodies
Pacific Blue™ anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis.
For test size, the suggested use of this reagent for immunofluorescent staining is 5 μl per million cells or 5 μl per 100 μl of whole blood.
For μg sizes, the suggested use of this reagent for immunofluorescent staining is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood.
It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: OKT4
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR