Anti-CD4 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: RPA-T4]

Supplier: Biolegend
300521 300524
CA300521-BLEA 427.67 CAD
CA300521-BL CA300524-BL
Anti-CD4 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: RPA-T4]
Antibodies
Pacific Blue™ anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Cynomolgus; Apps: FC; Size: 25 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 2.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Chimpanzee;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR